Tisk…

3-Amino-5-methoxypyridine 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD41913-250MGEA 428 CZK
BLDPBD41913-250MG BLDPBD41913-10G BLDPBD41913-1G BLDPBD41913-25G BLDPBD41913-5G
3-Amino-5-methoxypyridine 95%
3-Amino-5-methoxypyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₆H₈N₂O
MW: 124,1400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09701369
CAS číslo: 64436-92-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR