2-Chloro-3-nitrobenzaldehyde 95%
|
|
BLDPBD4194-10GEA
9640
CZK
BLDPBD4194-10G
BLDPBD4194-5G
BLDPBD4194-1G
BLDPBD4194-25G
BLDPBD4194-100MG
BLDPBD4194-250MG
2-Chloro-3-nitrobenzaldehyde 95%
2-Chloro-3-nitrobenzaldehyde
Storage condition: Inert atmosphere, 2-8°C
Vzorec:
C₇H₄ClNO₃ MW: 185,5700 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08236804 CAS číslo: 58755-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...