5-Cyano-2-fluorophenylboronic acid 97%
|
|
BLDPBD4282-5GEA
2570
CZK
BLDPBD4282-5G
BLDPBD4282-250MG
BLDPBD4282-100MG
BLDPBD4282-25G
BLDPBD4282-1G
5-Cyano-2-fluorophenylboronic acid 97%
5-Cyano-2-fluorophenylboronic acid
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Vzorec:
C₇H₅BFNO₂ MW: 164,9300 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03095147 CAS číslo: 468718-30-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...