Tisk…

5-Bromo-1-methyl-1H-pyrazolo[3,4-c]pyridine 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD446597-100MGEA 1320 CZK
BLDPBD446597-100MG BLDPBD446597-250MG BLDPBD446597-1G
5-Bromo-1-methyl-1H-pyrazolo[3,4-c]pyridine 98%
5-Bromo-1-methyl-1H-pyrazolo[3,4-c]pyridine

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₇H₆BrN₃
MW: 212,047 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22380195
CAS číslo: 1337880-46-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR