Tisk…

2-Methylimidazo[1,2-a]pyridin-6-amine 97%

BLDPBD447030-100MGEA 1340 CZK
BLDPBD447030-100MG BLDPBD447030-250MG
2-Methylimidazo[1,2-a]pyridin-6-amine 97%
2-Methylimidazo[1,2-a]pyridin-6-amine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₈H₉N₃
MW: 147,177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09991758
CAS číslo: 860258-05-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR