Tisk…

4-Fluoro-3-formylphenylboronic acid 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD4479-1GEA 200 CZK
BLDPBD4479-1G BLDPBD4479-5G BLDPBD4479-10G BLDPBD4479-250MG BLDPBD4479-100G BLDPBD4479-25G
4-Fluoro-3-formylphenylboronic acid 97%
4-Fluoro-3-formylphenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₇H₆BFO₃
MW: 167,93 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02093074
CAS číslo: 374538-01-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR