Tisk…

2-Chloro-6-methylnicotinic acid 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD4588-5GEA 371 CZK
BLDPBD4588-5G BLDPBD4588-25G BLDPBD4588-1G BLDPBD4588-10G BLDPBD4588-100G
2-Chloro-6-methylnicotinic acid 98%
2-Chloro-6-methylnicotinic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₇H₆ClNO₂
MW: 171,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00134165
CAS číslo: 30529-70-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR