(R)-(+)-2-Benzyloxypropionic acid 95%
Dodavatel: BLD PHARMATECH GMBH
|
|
BLDPBD47056-5GEA
1140
CZK
BLDPBD47056-5G
BLDPBD47056-10G
BLDPBD47056-1G
BLDPBD47056-25G
BLDPBD47056-250MG
BLDPBD47056-100MG
(R)-(+)-2-Benzyloxypropionic acid 95%
(R)-(+)-2-Benzyloxypropionic acid
Storage condition: Sealed in dry, Room Temperature
Vzorec:
C₁₀H₁₂O₃ MW: 180,20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06799065 CAS číslo: 100836-85-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...