Tisk…

5-Chloro-2-nitro-p-toluidine 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD4773-5GEA 1320 CZK
BLDPBD4773-5G BLDPBD4773-25G BLDPBD4773-1G
5-Chloro-2-nitro-p-toluidine 97%
5-Chloro-2-nitro-p-toluidine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₇H₇ClN₂O₂
MW: 186,60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00034066
CAS číslo: 7149-80-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR