Tisk…

3,4-Dihydroxybenzylamine hydrobromide 97%

BLDPBD5081-250MGEA 1200 CZK
BLDPBD5081-250MG BLDPBD5081-1G
3,4-Dihydroxybenzylamine hydrobromide 97%
3,4-Dihydroxybenzylamine hydrobromide

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₇H₁₀BrNO₂
MW: 220,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012859
CAS číslo: 16290-26-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR