Tisk…

Phenylmethanesulphonamide 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD5093-250MGEA 171 CZK
BLDPBD5093-250MG BLDPBD5093-100G BLDPBD5093-5G BLDPBD5093-1G BLDPBD5093-25G
Phenylmethanesulphonamide 95%
Phenylmethanesulphonamide

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₇H₉NO₂S
MW: 171,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041893
CAS číslo: 4563-33-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR