Tisk…

1-(4-Iodophenyl)-1H-pyrazole 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD51183-5GEA 9690 CZK
BLDPBD51183-5G BLDPBD51183-1G BLDPBD51183-250MG
1-(4-Iodophenyl)-1H-pyrazole 98%
1-(4-Iodophenyl)-1H-pyrazole

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₉H₇IN₂
MW: 270,0700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02681941
CAS číslo: 368869-86-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR