Tisk…

(3-Chloro-4-methoxyphenyl)methanamine hydrochloride 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD5148-25GEA 2740 CZK
BLDPBD5148-0.025G BLDPBD5148-25G BLDPBD5148-5G BLDPBD5148-10G
(3-Chloro-4-methoxyphenyl)methanamine hydrochloride 98%
(3-Chloro-4-methoxyphenyl)methanamine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₈H₁₁Cl₂NO
MW: 208,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01211460
CAS číslo: 41965-95-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR