N-Methyl-1-(3-nitrophenyl)methanamine 98%
|
|
BLDPBD5201-1GEA
428
CZK
BLDPBD5201-1G
N-Methyl-1-(3-nitrophenyl)methanamine 98%
N-Methyl-1-(3-nitrophenyl)methanamine
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Vzorec:
C₈H₁₀N₂O₂ MW: 166,18 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD04115408 CAS číslo: 19499-61-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...