Tisk…

2-Butyl-5-chloroimidazole-4-carbaldehyde 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD5283-1000GEA 5330 CZK
BLDPBD5283-1000G BLDPBD5283-500G BLDPBD5283-0.5G BLDPBD5283-0.025G BLDPBD5283-25G BLDPBD5283-10G BLDPBD5283-100G
2-Butyl-5-chloroimidazole-4-carbaldehyde 97%
2-Butyl-5-chloroimidazole-4-carbaldehyde

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₈H₁₁ClN₂O
MW: 186,6387 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01934396
CAS číslo: 83857-96-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR