Tisk…

1,3-Dicyclohexyl-2-thiourea 98%

BLDPBD54028-25GEA 1290 CZK
BLDPBD54028-25G BLDPBD54028-100G BLDPBD54028-0.025G BLDPBD54028-5G
1,3-Dicyclohexyl-2-thiourea 98%
1,3-Dicyclohexyl-2-thiourea

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₃H₂₄N₂S
MW: 240,41 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00792562
CAS číslo: 1212-29-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR