Tisk…

1,3-Diphenoxybenzene 95%

BLDPBD54228-1GEA 513 CZK
BLDPBD54228-1G BLDPBD54228-25G BLDPBD54228-5G
1,3-Diphenoxybenzene 95%
1,3-Diphenoxybenzene

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₈H₁₄O₂
MW: 262,31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003036
CAS číslo: 3379-38-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR