1-[3,5-Bis(trifluoromethyl)phenyl]-2,5-dimethylpyrrole 97%
|
|
BLDPBD55492-25GEA
14540
CZK
BLDPBD55492-25G
BLDPBD55492-1G
BLDPBD55492-5G
1-[3,5-Bis(trifluoromethyl)phenyl]-2,5-dimethylpyrrole 97%
1-[3,5-Bis(trifluoromethyl)phenyl]-2,5-dimethylpyrrole
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Vzorec:
C₁₄H₁₁F₆N MW: 307,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00084902 CAS číslo: 175205-51-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...