Tisk…

3-Bromo-5-(trifluoromethyl)benzoic acid 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD5599-500GEA 16450 CZK
BLDPBD5599-500G BLDPBD5599-5G BLDPBD5599-25G BLDPBD5599-10G BLDPBD5599-1G BLDPBD5599-100G
3-Bromo-5-(trifluoromethyl)benzoic acid 97%
3-Bromo-5-(trifluoromethyl)benzoic acid

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₈H₄BrF₃O₂
MW: 269,02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03412186
CAS číslo: 328-67-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR