1-Acetylpyrene 98+%
|
|
BLDPBD56239-5GEA
4170
CZK
BLDPBD56239-5G
BLDPBD56239-1G
BLDPBD56239-10G
1-Acetylpyrene 98+%
1-Acetylpyrene
Storage condition: Sealed in dry, Room Temperature
Vzorec:
C₁₈H₁₂O MW: 244,29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00021663 CAS číslo: 3264-21-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...