Tisk…

Decane-1,10-diamine, N-BOC protected 95%

BLDPBD56873-5GEA 12890 CZK
BLDPBD56873-5G BLDPBD56873-25G BLDPBD56873-250MG BLDPBD56873-1G
Decane-1,10-diamine, N-BOC protected 95%
Decane-1,10-diamine, N-BOC protected

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₁₅H₃₂N₂O₂
MW: 272,427 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02094501
CAS číslo: 216961-61-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR