Tisk…

5-Chloro-2-methoxynicotinonitrile 97%

BLDPBD572908-1GEA 4510 CZK
BLDPBD572908-1G BLDPBD572908-100MG BLDPBD572908-250MG
5-Chloro-2-methoxynicotinonitrile 97%
5-Chloro-2-methoxynicotinonitrile
Vzorec: C₇H₅ClN₂O
MW: 168,58 g/mol
MDL Number: MFCD17214212
CAS číslo: 1256812-94-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR