Tisk…

3,5-Dibromo-4-methylbenzoic acid 97%

BLDPBD5806-1GEA 1060 CZK
BLDPBD5806-1G BLDPBD5806-25G BLDPBD5806-250MG BLDPBD5806-5G
3,5-Dibromo-4-methylbenzoic acid 97%
3,5-Dibromo-4-methylbenzoic acid

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₈H₆Br₂O₂
MW: 293,94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017544
CAS číslo: 67973-32-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR