Tisk…

Dibenzo[b,d]furan-4-ol 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD582448-25GEA 4940 CZK
BLDPBD582448-25G BLDPBD582448-5G BLDPBD582448-1G
Dibenzo[b,d]furan-4-ol 97%
Dibenzo[b,d]furan-4-ol

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₂H₈O₂
MW: 184,191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00092287
CAS číslo: 19261-06-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR