Tisk…

2-Fluoro-4'-chloroacetophenone 95+%

BLDPBD5874-250MGEA 2710 CZK
BLDPBD5874-250MG BLDPBD5874-100MG BLDPBD5874-1G
2-Fluoro-4'-chloroacetophenone 95+%
2-Fluoro-4'-chloroacetophenone

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₈H₆ClFO
MW: 172,5840 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03412196
CAS číslo: 329-78-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR