Methyl-1H-indazole-7-carboxylate 97%
Dodavatel: BLD PHARMATECH GMBH
|
|
BLDPBD58805-1GEA
599
CZK
BLDPBD58805-1G
BLDPBD58805-10G
BLDPBD58805-25G
BLDPBD58805-5G
Methyl-1H-indazole-7-carboxylate 97%
Methyl-1H-indazole-7-carboxylate
Storage condition: Sealed in dry, Room Temperature
Vzorec:
C₉H₈N₂O₂ MW: 176,1720 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07371608 CAS číslo: 755752-82-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...