Tisk…

3-Bromo-1H-pyrazolo[3,4-b]pyridine 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD58924-25GEA 7440 CZK
BLDPBD58924-25G BLDPBD58924-1G BLDPBD58924-10G BLDPBD58924-5G
3-Bromo-1H-pyrazolo[3,4-b]pyridine 97%
3-Bromo-1H-pyrazolo[3,4-b]pyridine

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₆H₄BrN₃
MW: 198,0200 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08669476
CAS číslo: 68618-36-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR