Tisk…

Methyl-1H-pyrrole-3-carboxylate 97%

BLDPBD58954-25GEA 6870 CZK
BLDPBD58954-25G BLDPBD58954-0.025G BLDPBD58954-5G BLDPBD58954-100G BLDPBD58954-1G BLDPBD58954-10G
Methyl-1H-pyrrole-3-carboxylate 97%
Methyl-1H-pyrrole-3-carboxylate

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₆H₇NO₂
MW: 125,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00967074
CAS číslo: 2703-17-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR