Tisk…

2-Methyl-6-(trifluoromethyl)nicotinic acid 95%

BLDPBD5949-250MGEA 684 CZK
BLDPBD5949-250MG BLDPBD5949-1G BLDPBD5949-10G BLDPBD5949-5G
2-Methyl-6-(trifluoromethyl)nicotinic acid 95%
2-Methyl-6-(trifluoromethyl)nicotinic acid

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₈H₆F₃NO₂
MW: 205,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01311990
CAS číslo: 261635-93-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR