Tisk…

1-Methyl-4-nitro-1H-indazole 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD59499-1GEA 1370 CZK
BLDPBD59499-1G BLDPBD59499-250MG
1-Methyl-4-nitro-1H-indazole 98%
1-Methyl-4-nitro-1H-indazole

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₈H₇N₃O₂
MW: 177,1600 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09607929
CAS číslo: 26120-43-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR