Tisk…

2,3-Dichloro-5-(trichloromethyl)pyridine 95%

BLDPBD60577-1GEA 3450 CZK
BLDPBD60577-1G BLDPBD60577-250MG
2,3-Dichloro-5-(trichloromethyl)pyridine 95%
2,3-Dichloro-5-(trichloromethyl)pyridine

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₆H₂Cl₅N
MW: 265,3518 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03701543
CAS číslo: 69045-83-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR