Tisk…

1-(2-Bromoethoxy)-2-ethoxybenzene 98%

BLDPBD61279-5GEA 485 CZK
BLDPBD61279-5G BLDPBD61279-100G BLDPBD61279-10G BLDPBD61279-25G
1-(2-Bromoethoxy)-2-ethoxybenzene 98%
1-(2-Bromoethoxy)-2-ethoxybenzene

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₀H₁₃BrO₂
MW: 245,12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02030483
CAS číslo: 3259-03-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR