Tisk…

2,4-Diamino-5-fluoroquinazoline 98%

BLDPBD6154-5GEA 5790 CZK
BLDPBD6154-5G BLDPBD6154-1G BLDPBD6154-250MG
2,4-Diamino-5-fluoroquinazoline 98%
2,4-Diamino-5-fluoroquinazoline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₈H₇FN₄
MW: 178,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00221785
CAS číslo: 119584-70-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR