Tisk…

(3,5-Dichlorophenyl)acetic acid 97%

BLDPBD61620-25GEA 2800 CZK
BLDPBD61620-25G BLDPBD61620-10G BLDPBD61620-1G BLDPBD61620-5G BLDPBD61620-100G BLDPBD61620-500G
(3,5-Dichlorophenyl)acetic acid 97%
(3,5-Dichlorophenyl)acetic acid

Storage condition: Sealed in dry, Store in freezer, under -20°C

Vzorec: C₈H₆Cl₂O₂
MW: 205,03 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01764671
CAS číslo: 51719-65-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR