Tisk…

4,4'-Dicyanobenzhydrol 97%

BLDPBD6221-25GEA 6590 CZK
BLDPBD6221-25G BLDPBD6221-1G BLDPBD6221-5G BLDPBD6221-250MG
4,4'-Dicyanobenzhydrol 97%
4,4'-Dicyanobenzhydrol

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₅H₁₀N₂O
MW: 234,2500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07367966
CAS číslo: 134521-16-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR