1-Naphthyl boronic acid neopentyl glycol ester 97%
Dodavatel: BLD PHARMATECH GMBH
|
|
BLDPBD626281-1GEA
884
CZK
BLDPBD626281-1G
BLDPBD626281-250MG
1-Naphthyl boronic acid neopentyl glycol ester 97%
1-Naphthyl boronic acid neopentyl glycol ester
Storage condition: Sealed in dry, 2-8°C
Vzorec:
C₁₅H₁₇BO₂ MW: 240,11 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01318167 CAS číslo: 22871-77-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...