Tisk…

5-Indoleboronic acid 97%

BLDPBD6264-1GEA 713 CZK
BLDPBD6264-1G BLDPBD6264-250MG BLDPBD6264-0.025G BLDPBD6264-10G BLDPBD6264-5G BLDPBD6264-25G
5-Indoleboronic acid 97%
5-Indoleboronic acid

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₈H₈BNO₂
MW: 160,97 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01319013
CAS číslo: 144104-59-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR