Tisk…

4-Bromo-3-methylbenzamide 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD6274-5GEA 656 CZK
BLDPBD6274-5G BLDPBD6274-25G BLDPBD6274-1G
4-Bromo-3-methylbenzamide 98%
4-Bromo-3-methylbenzamide

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₈H₈BrNO
MW: 214,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00672914
CAS číslo: 170229-98-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR