Dodecacarbonyltetrarhodium 98%
|
|
BLDPBD634044-250MGEA
15250
CZK
BLDPBD634044-250MG
BLDPBD634044-1G
BLDPBD634044-100MG
Dodecacarbonyltetrarhodium 98%
Dodecacarbonyltetrarhodium
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Vzorec:
C₁₂O₁₂Rh₄ MW: 747,743 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00016310 CAS číslo: 19584-30-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...