(R)-b-Amino-4-methyl-benzeneethanol 95%
|
|
BLDPBD635144-250MGEA
7470
CZK
BLDPBD635144-250MG
BLDPBD635144-100MG
(R)-b-Amino-4-methyl-benzeneethanol 95%
(R)-b-Amino-4-methyl-benzeneethanol
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Vzorec:
C₉H₁₃NO MW: 151,206 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09253779 CAS číslo: 639054-51-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...