Tisk…

5-(4-Methylphenyl)-1H-tetrazole 98%

BLDPBD6370-5GEA 3080 CZK
BLDPBD6370-5G BLDPBD6370-1G BLDPBD6370-25G
5-(4-Methylphenyl)-1H-tetrazole 98%
5-(4-Methylphenyl)-1H-tetrazole

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₈H₈N₄
MW: 160,18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01318164
CAS číslo: 24994-04-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR