Tisk…

4-Hydroxy-2-methylbenzaldehyde 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD6382-10GEA 4140 CZK
BLDPBD6382-10G BLDPBD6382-5G BLDPBD6382-1G BLDPBD6382-250MG
4-Hydroxy-2-methylbenzaldehyde 97%
4-Hydroxy-2-methylbenzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₈H₈O₂
MW: 136,1479 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04037443
CAS číslo: 41438-18-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR