Tisk…

4-(Methylsulphonyl)benzoic acid 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD6425-100GEA 1690 CZK
BLDPBD6425-100G BLDPBD6425-5G BLDPBD6425-10G BLDPBD6425-500G BLDPBD6425-25G
4-(Methylsulphonyl)benzoic acid 97%
4-(Methylsulphonyl)benzoic acid

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₈H₈O₄S
MW: 200,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007564
CAS číslo: 4052-30-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR