Tisk…

3-Bromo-5-methylanisole 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD6458-100GEA 6130 CZK
BLDPBD6458-100G BLDPBD6458-10G BLDPBD6458-1G BLDPBD6458-25G BLDPBD6458-5G BLDPBD6458-500G
3-Bromo-5-methylanisole 97%
3-Bromo-5-methylanisole

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₈H₉BrO
MW: 201,0600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061916
CAS číslo: 29578-83-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR