Tisk…

2,3,6-Trichloroquinoxaline 98%

BLDPBD65238-5GEA 12570 CZK
BLDPBD65238-5G BLDPBD65238-100MG BLDPBD65238-250MG BLDPBD65238-1G
2,3,6-Trichloroquinoxaline 98%
2,3,6-Trichloroquinoxaline

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₈H₃Cl₃N₂
MW: 233,4800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006721
CAS číslo: 2958-87-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR