Tisk…

3-(Aminomethyl)benzoic acid hydrochloride 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD6525-10GEA 3940 CZK
BLDPBD6525-10G BLDPBD6525-250MG BLDPBD6525-5G BLDPBD6525-25G BLDPBD6525-100MG BLDPBD6525-1G
3-(Aminomethyl)benzoic acid hydrochloride 97%
3-(Aminomethyl)benzoic acid hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₈H₁₀ClNO₂
MW: 187,62 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03791117
CAS číslo: 876-03-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR