Tisk…

4-Ethoxy-3-(trifluoromethyl)phenylboronic acid 98%

BLDPBD6574-1GEA 1690 CZK
BLDPBD6574-1G BLDPBD6574-250MG
4-Ethoxy-3-(trifluoromethyl)phenylboronic acid 98%
4-Ethoxy-3-(trifluoromethyl)phenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₉H₁₀BF₃O₃
MW: 233,98 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07363789
CAS číslo: 871329-83-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR