Tisk…

5-Ethyl-2,3-dimethylpyrazine 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD65837-1GEA 485 CZK
BLDPBD65837-1G
5-Ethyl-2,3-dimethylpyrazine 95%
5-Ethyl-2,3-dimethylpyrazine
Vzorec: C₈H₁₂N₂
MW: 136,1900 g/mol
MDL Number: MFCD04038059
CAS číslo: 15707-34-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR