Tisk…

5,6-Dimethoxybenzimidazole 98%

BLDPBD6625-1GEA 2660 CZK
BLDPBD6625-1G BLDPBD6625-250MG BLDPBD6625-5G
5,6-Dimethoxybenzimidazole 98%
5,6-Dimethoxybenzimidazole

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Vzorec: C₉H₁₀N₂O₂
MW: 178,19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00612461
CAS číslo: 72721-02-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR