Tisk…

2',4'-Dihydroxy-2-phenylacetophenone 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD66913-250MGEA 257 CZK
BLDPBD66913-250MG BLDPBD66913-1G BLDPBD66913-5G
2',4'-Dihydroxy-2-phenylacetophenone 98%
2',4'-Dihydroxy-2-phenylacetophenone

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₄H₁₂O₃
MW: 228,2433 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00043754
CAS číslo: 3669-41-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR